Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03682545_s1
          See other 1700008J07RIK GE Assays ›
          Gene Symbol:
          1700008J07Rik
          Gene Name:
          RIKEN cDNA 1700008J07 gene
          Gene Aliases:
          AW048743
          Chromosome Location:
          Chr.7: 127510438 - 127512869 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03682545_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700008J07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024331.1 1 1949 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB391151.2 - 1 174 Chr. 7 - 127510438 - 127511094 [-] on Build GRCm38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Snf2-related CREBBP activator protein Srcap B930091H02Rik,
          D030022P06Rik,
          F630004O05Rik
          Anti-sense Chr. 7 - 127511983 - 127561219 on NCBI Build GRCm38
          Downstream small nucleolar RNA,
          H/ACA box 30
          Snora30 MBI-26 15.0kb Anti-sense Chr. 7 - 127527897 - 127528003 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004330.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114870 NR_024331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114870 AK082635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154235 BC085144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154235 AK142089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154235 AK051146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154235 BC049754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154235 AK131716.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173676 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114870 NR_024331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114870 AK082635.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173676 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114870 NR_024331.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114870 AK082635.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102865 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114932 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06503351 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568259 - -
          Affymetrix HT MG-430 PM Array Plate 1428732_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441221_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568258 - -
          Mouse Genome 430 2.0 Array 1428732_at - -
          Mouse Genome 430 2.0 Array 1441221_PM_at - -
          Mouse Genome 430 2.0 Array 1441221_at - -
          Mouse Genome 430 2.0 Array 1428732_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700036367.mm.1 TC0700004330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036367.mm.1 NR_024331.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036367.mm.1 NR_024331.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036366.mm.1 TC0700004330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036366.mm.1 NR_024331.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036366.mm.1 NR_024331.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036368.mm.1 TC0700004330.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036368.mm.1 NR_024331.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036368.mm.1 NR_024331.1 Refseq
          Mouse Expression Set 430 1428732_at - -
          Mouse Expression Set 430 1441221_PM_at - -
          Mouse Expression Set 430 1428732_PM_at - -
          Mouse Expression Set 430 1441221_at - -
          Affymetrix HT Mouse Genome 430B Array 1428732_at - -
          Affymetrix HT Mouse Genome 430B Array 1441221_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428732_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441221_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 647017 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106370671 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222691 - -
          Illumina Sentrix mouse-6.v1 bead array 106370671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106370671 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2580171 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0