Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03682622_s1
          See other C030034I22RIK GE Assays ›
          Gene Symbol:
          C030034I22Rik
          Gene Name:
          RIKEN cDNA C030034I22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 69416447 - 69419193 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03682622_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C030034I22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026848.1 1 148 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE949423.1 - 1 168 Chr. 17 - 69416427 - 69416975 [+] on Build GRCm38 69
          GenBankmRNA BB182448.2 - 1 Chr. 17 - 69416225 - 69416873 [-] on Build GRCm38 69
          GenBankmRNA AI846356.1 - 1 168 Chr. 17 - 69416427 - 69416792 [+] on Build GRCm38 69
          GenBankmRNA BG694899.1 - 1 Chr. 17 - 69416282 - 69416966 [-] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA A930029G22 gene A930029G22Rik Anti-sense Chr. 17 - 69356361 - 69440118 on NCBI Build GRCm38
          Downstream RIKEN cDNA A330050F15 gene A330050F15Rik 18.4kb Sense Chr. 17 - 69437591 - 69489233 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001111.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133577 NR_026848.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133577 AK147961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133577 AK165105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112934 AI482189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112934 NR_026848.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112934 AK147961.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558284 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133577 NR_026848.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133577 AK147961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133577 AK165105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112934 AI482189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112934 NR_026848.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112934 AK147961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P778544 AK021121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P778544 NR_026848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P778544 AK147961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P778544 AK165105.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13247417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31323858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10446588 - -
          Affymetrix HT MG-430 PM Array Plate 1435909_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10446587 - -
          Mouse Genome 430 2.0 Array 1435909_PM_at - -
          Mouse Genome 430 2.0 Array 1435909_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77533_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77533_at - -
          Affymetrix Mouse Clariom S Array PSR1700010465.mm.1 TC1700001111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010465.mm.1 NR_026848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010465.mm.1 NR_026848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010461.mm.1 TC1700001111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010461.mm.1 NR_026848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010461.mm.1 NR_026848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010464.mm.1 TC1700001111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010464.mm.1 NR_026848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010464.mm.1 NR_026848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010462.mm.1 TC1700001111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010462.mm.1 NR_026848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010462.mm.1 NR_026848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010463.mm.1 TC1700001111.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700010463.mm.1 NR_026848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700010463.mm.1 NR_026848.1 Refseq
          Mouse Expression Set 430 1435909_at - -
          Mouse Expression Set 430 1435909_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435909_at - -
          Affymetrix HT Mouse Genome 430B Array 1435909_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 527241 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0