Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159349.1 | NP_001152821.1 | 1 - 2 | 211 | 89 | |
NM_027226.4 | NP_081502.2 | 1 - 2 | 211 | 89 | ||
GenBank mRNA | AJ344096.1 | - | 1 - 2 | 95 | 89 | |
AK008130.1 | - | 1 - 2 | 183 | 89 | ||
AK014447.1 | - | 1 - 2 | 182 | 89 | ||
AK029785.1 | - | 1 - 2 | 156 | 89 | ||
AK050448.1 | - | 1 - 2 | 170 | 89 | ||
AK050479.1 | - | 1 - 2 | 183 | 89 | ||
AK139638.1 | - | 1 - 2 | 207 | 89 | ||
AK157602.1 | - | 1 - 2 | 183 | 89 | ||
AK161252.1 | - | 1 - 2 | 189 | 89 | ||
AK165633.1 | - | 1 - 2 | 165 | 89 | ||
AK166032.1 | - | 1 - 2 | 183 | 89 | ||
AK170511.1 | - | 1 - 2 | 183 | 89 | ||
BC010204.1 | - | 1 - 2 | 185 | 3158224 | 89 | |
BC028253.1 | - | 1 - 2 | 168 | 3983533 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |