Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929236_s1
          See other 1700008A07RIK GE Assays ›
          Gene Symbol:
          1700008A07Rik
          Gene Name:
          RIKEN cDNA 1700008A07 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929236_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700008A07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005750.1 - 1 652 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dopachrome tautomerase Dct DT,
          TRP-2,
          TRP2,
          Tyrp-2,
          Tyrp2,
          slaty,
          slt
          -36.2kb Sense Chr. 14 - 118012790 - 118052323 on NCBI Build GRCm38
          Downstream TDP-glucose 4,6-dehydratase Tgds 2610017J16Rik,
          2610025M23Rik,
          AI648925
          22.6kb Sense Chr. 14 - 118111911 - 118132765 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_52_P43168 AK005750.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P11186475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11161322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11453265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11735668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11735670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11307065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11670567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11735667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29936553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11453249 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 354884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-kqswr:80/100.66.130.62:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0