Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929272_s1
          See other 1700010B09RIK GE Assays ›
          Gene Symbol:
          1700010B09Rik
          Gene Name:
          RIKEN cDNA 1700010B09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929272_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700010B09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005812.1 - 1 50 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF023120.1 - 1 Chr. 14 - 79301011 - 79301438 [-] on Build GRCm38 64
          GenBankmRNA AA197429.1 - 1 Chr. 14 - 79300965 - 79301435 [-] on Build GRCm38 64
          GenBankmRNA AI646229.1 - 1 Chr. 14 - 79301051 - 79301435 [-] on Build GRCm38 64
          GenBankmRNA BF019893.1 - 1 278 Chr. 14 - 79300920 - 79301363 [+] on Build GRCm38 64
          GenBankmRNA BX519735.1 - 1 Chr. 14 - 79300902 - 79301435 [-] on Build GRCm38 64
          GenBankmRNA BE847359.1 - 1 Chr. 14 - 79300984 - 79301438 [-] on Build GRCm38 64
          GenBankmRNA BU961387.1 - 54 Chr. - - [-] on Build 64
          GenBankmRNA BE332894.1 - 1 Chr. 14 - 79301084 - 79301438 [-] on Build GRCm38 64
          GenBankmRNA BY705965.1 - 50 Chr. - - [-] on Build 64
          GenBankmRNA BE457695.1 - 1 322 Chr. 14 - 79300876 - 79301371 [+] on Build GRCm38 64
          GenBankmRNA BU961580.1 - 54 Chr. - - [-] on Build 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream regulator of cell cycle Rgcc 1190002H23Rik,
          Rgc-32,
          Rgc32
          Anti-sense Chr. 14 - 79288750 - 79301635 on NCBI Build GRCm38
          Downstream N(alpha)-acetyltransferase 16,
          NatA auxiliary subunit
          Naa16 1300019C06Rik,
          Narg1l
          32.5kb Anti-sense Chr. 14 - 79334507 - 79390777 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440159_PM_at - -
          Mouse Genome 430 2.0 Array 1440159_at - -
          Mouse Genome 430 2.0 Array 1440159_PM_at - -
          Mouse Expression Set 430 1440159_at - -
          Mouse Expression Set 430 1440159_PM_at - -
          Murine Genome U74v2 Set 166642_PM_at - -
          Murine Genome U74v2 Set 166642_at - -
          Affymetrix HT Mouse Genome 430B Array 1440159_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440159_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 592676 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102630088 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0