Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929449_g1
          See other 1700019P21RIK GE Assays ›
          Gene Symbol:
          1700019P21Rik
          Gene Name:
          RIKEN cDNA 1700019P21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 138876272 - 138876965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929449_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700019P21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006139.1 - 1 - 2 37 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU936470.1 - 1 - 2 37 Chr. 1 - 138876272 - 138876950 [-] on Build GRCm38 63
          GenBankmRNA BY706245.1 - 1 - 2 36 Chr. 1 - 138876274 - 138876950 [-] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 2310009B15 gene 2310009B15Rik -19.4kb Sense Chr. 1 - 138851979 - 138856854 on NCBI Build GRCm38
          Downstream DENN/MADD domain containing 1B Dennd1b 4632404N19Rik,
          4930467M19Rik,
          6820401H01Rik,
          F730008N07Rik
          86.5kb Anti-sense Chr. 1 - 138963446 - 139176043 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899122 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039518 AK006139.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01077908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01014859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00814458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01014860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00257707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00806113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01014856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00257703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00806114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01077909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00987252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00257701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00257702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00058139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00058140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01014857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20659706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1447710_PM_at - -
          Mouse Genome 430 2.0 Array 1447710_PM_at - -
          Mouse Genome 430 2.0 Array 1447710_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75546_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75546_at - -
          Mouse Expression Set 430 1447710_at - -
          Mouse Expression Set 430 1447710_PM_at - -
          Murine Genome U74v2 Set 167605_PM_i_at - -
          Murine Genome U74v2 Set 167605_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1447710_at - -
          Affymetrix HT Mouse Genome 430B Array 1447710_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106040411 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0