Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929521_s1
          See other 1700027M17RIK GE Assays ›
          Gene Symbol:
          1700027M17Rik
          Gene Name:
          RIKEN cDNA 1700027M17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929521_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700027M17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006436.1 - 1 46 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 9376 Gm9376 EG668814 -11.5kb Sense Chr. 14 - 118267158 - 118267779 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700044C05 gene 1700044C05Rik 86.3kb Sense Chr. 14 - 118365878 - 118398165 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11749969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11654153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11576598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11311777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11151768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11576595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11654152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11219023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29937735 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 663858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-64bcc5cc5d-xqgf4:80/100.66.130.62:80.
          git-commit: fd21aaec18e3d7b1379e340bd1771dde3ea5c6fc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0