Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929549_g1
          See other 1700031P21RIK GE Assays ›
          Gene Symbol:
          1700031P21Rik
          Gene Name:
          RIKEN cDNA 1700031P21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 52599984 - 52602322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929549_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700031P21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045910.1 1 - 2 144 100
          GenBank mRNA AK006587.1 - 1 - 2 145 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF105345.1 - 1 - 2 128 Chr. 12 - 52601829 - 52602306 [-] on Build GRCm38 100
          GenBankmRNA CA494505.1 - 144 Chr. - - [-] on Build 100
          GenBankmRNA BY706590.1 - 1 - 2 140 Chr. 12 - 52599984 - 52602318 [-] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Rho GTPase activating protein 5 Arhgap5 p190-B,
          p190B
          -30.0kb Anti-sense Chr. 12 - 52503771 - 52571978 on NCBI Build GRCm38
          Downstream A kinase (PRKA) anchor protein 6 Akap6 AI482140,
          Akapalpha,
          Akapbeta,
          PRKA
          96.7kb Anti-sense Chr. 12 - 52699016 - 53155568 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001733.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171312 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09973723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10156145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09856051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09828839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10155950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09863140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10414626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10306445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28435812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200011943.mm.1 TC1200001733.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011943.mm.1 NR_045910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011943.mm.1 NR_045910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011937.mm.1 TC1200001733.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011937.mm.1 NR_045910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011937.mm.1 NR_045910.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011941.mm.1 TC1200001733.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011941.mm.1 NR_045910.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011941.mm.1 NR_045910.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631418 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101500270 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0