Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929550_g1
          See other R3HCC1L GE Assays ›
          Gene Symbol:
          R3hcc1l
          Gene Name:
          R3H domain and coiled-coil containing 1 like
          Gene Aliases:
          1700036B12Rik, AI450607, C10orf28, D19Ertd386e, GIDRP86, Gidrp88
          Chromosome Location:
          Chr.19: 42518312 - 42592256 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929550_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: R3hcc1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006610.1 - 1 - 2 139 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA494603.1 - 105 Chr. - - [-] on Build 92
          GenBankmRNA BU938526.1 - 105 Chr. - - [-] on Build 92
          GenBankmRNA CA463283.1 - 1 - 2 114 Chr. 19 - 42536850 - 42563083 [-] on Build GRCm38 92
          GenBankmRNA BU938197.1 - 105 Chr. - - [-] on Build 92
          GenBankmRNA AV047259.3 - 336 Chr. - - [-] on Build 92
          GenBankmRNA CA463393.1 - 1 - 2 115 Chr. 19 - 42562499 - 42563083 [-] on Build GRCm38 92
          GenBankmRNA CN836157.1 - 115 Chr. - - [-] on Build 92
          GenBankmRNA BU962299.1 - 105 Chr. - - [-] on Build 92
          GenBankmRNA BY091223.1 - 1 - 2 146 Chr. 19 - 42562598 - 42563114 [-] on Build GRCm38 92
          GenBankmRNA CR520889.1 - 1 - 2 Chr. 19 - 42562502 - 42563046 [+] on Build GRCm38 92
          GenBankmRNA BU939654.1 - 115 Chr. - - [-] on Build 92
          GenBankmRNA AV207571.2 - 1 - 2 133 Chr. 19 - 42562525 - 42563081 [-] on Build GRCm38 92
          GenBankmRNA BY706609.1 - 1 - 2 139 Chr. 19 - 42562504 - 42563108 [-] on Build GRCm38 92
          GenBankmRNA BU962179.1 - 115 Chr. - - [-] on Build 92
          GenBankmRNA BY455785.1 - 1 - 2 134 Chr. 19 - 42562504 - 42563103 [-] on Build GRCm38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cartilage acidic protein 1 Crtac1 2810454P21Rik,
          AW047536,
          Crtac1B,
          Lotus
          -130.7kb Sense Chr. 19 - 42283036 - 42431820 on NCBI Build GRCm38
          Downstream lysyl oxidase-like 4 Loxl4 4833426I20Rik,
          Loxc
          29.2kb Sense Chr. 19 - 42592279 - 42612906 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 794380 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0