Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929599_s1
          See other 1700042G15RIK GE Assays ›
          Gene Symbol:
          1700042G15Rik
          Gene Name:
          RIKEN cDNA 1700042G15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 57359792 - 57364292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929599_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700042G15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038178.1 3 1620 108
          NR_038179.1 3 1573 108
          GenBank mRNA AK006686.1 - 1 249 108
          AK133010.1 - 3 1575 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV270740.2 - 1 192 Chr. 4 - 57359797 - 57360392 [-] on Build GRCm38 108
          GenBankmRNA BY706675.1 - 1 241 Chr. 4 - 57360056 - 57360442 [-] on Build GRCm38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein tyrosine phosphatase,
          non-receptor type 3
          Ptpn3 9530011I20Rik,
          PTP-H1,
          PTPCL
          -21.0kb Sense Chr. 4 - 57190841 - 57339046 on NCBI Build GRCm38
          Downstream paralemmin 2 Palm2 53-H12,
          B130066I24,
          R75368,
          pEN70
          207.8kb Anti-sense Chr. 4 - 57568248 - 57717146 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002721.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150598 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112065 AK133010.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03932044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03873298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03828580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03458623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03932041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03915912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03973873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03709521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03985935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03669169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03669172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03975138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03669173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04058354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03529665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03353854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22945025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22944982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400022021.mm.1 TC0400002721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022021.mm.1 NR_038178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022021.mm.1 NR_038178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022021.mm.1 NR_038179.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022021.mm.1 NR_038179.1 Refseq
          Murine Genome U74v2 Set 169687_at - -
          Murine Genome U74v2 Set 169687_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 445175 - -
          ABI Mouse Genome Survey Microarray 636463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix