Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929689_m1
          See other 1700063H06RIK GE Assays ›
          Gene Symbol:
          1700063H06Rik
          Gene Name:
          RIKEN cDNA 1700063H06 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929689_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700063H06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006870.1 - 1 - 2 144 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706855.1 - 1 - 2 143 Chr. 9 - 82260294 - 82263485 [+] on Build GRCm38 87
          GenBankmRNA CB273662.1 - 1 - 2 93 Chr. 9 - 82260348 - 82263449 [+] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream meiosis-specific,
          MEI4 homolog (S. cerevisiae)
          Mei4 4930486G11Rik -54.3kb Sense Chr. 9 - 81863647 - 82206007 on NCBI Build GRCm38
          Downstream interleukin-1 receptor-associated kinase 1 binding protein 1 Irak1bp1 4921528N06Rik,
          AI851240,
          Aabp3,
          Aip70,
          Simpl
          566.3kb Sense Chr. 9 - 82829806 - 82847688 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08032874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08172635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07605514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08032888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08032902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08141647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26677942 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 856242 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100070519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0