Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929740_s1
          See other 1700084K02RIK GE Assays ›
          Gene Symbol:
          1700084K02Rik
          Gene Name:
          RIKEN cDNA 1700084K02 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929740_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700084K02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006995.1 - 745 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV206122.1 - 1 162 Chr. 19 - 43940621 - 43940920 [+] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dynamin binding protein Dnmbp 2410003L07Rik,
          2410003M15Rik,
          TUBA
          -303.0 Anti-sense Chr. 19 - 43846815 - 43940318 on NCBI Build GRCm38
          Downstream carboxypeptidase N,
          polypeptide 1
          Cpn1 0610011F20Rik,
          CPN
          15.4kb Anti-sense Chr. 19 - 43956307 - 43986551 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P35072 AK006995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P35072 AK006995.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151203 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159514 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P35072 AK006995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P35072 AK006995.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159514 - -
          Whole Mouse Genome G4122A A_52_P35072 AK006995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P35072 AK006995.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129421 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14301000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14507715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14213434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14468112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14289315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14213433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14394392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14213442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14301003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32129568 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463346 - -
          Affymetrix HT MG-430 PM Array Plate 1433406_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463345 - -
          Mouse Genome 430 2.0 Array 1433405_PM_at - -
          Mouse Genome 430 2.0 Array 1433406_at - -
          Mouse Genome 430 2.0 Array 1433405_at - -
          Mouse Genome 430 2.0 Array 1433406_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 668408_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 668408_at - -
          Mouse Expression Set 430 1433405_at - -
          Mouse Expression Set 430 1433405_PM_at - -
          Mouse Expression Set 430 1433406_at - -
          Mouse Expression Set 430 1433406_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433406_at - -
          Affymetrix HT Mouse Genome 430B Array 1433405_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433406_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433405_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101340170 - -
          Illumina mouse-6 v1.0 expression beadchip 101340170 - -
          Illumina Sentrix mouse-6.v1 bead array 101340170 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340170 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0