Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10424422 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1440839_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1440839_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1440839_x_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1500019338.mm.1 |
TC1500002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500019338.mm.1 |
NR_040315.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500019338.mm.1 |
NR_108057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003277.mm.1 |
TC1500002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003277.mm.1 |
NR_040315.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003277.mm.1 |
NR_108057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003274.mm.1 |
TC1500002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003274.mm.1 |
NR_108057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003275.mm.1 |
TC1500002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003275.mm.1 |
NR_040315.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003275.mm.1 |
NR_108057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003276.mm.1 |
TC1500002331.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003276.mm.1 |
NR_040315.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003276.mm.1 |
NR_108057.1 |
Refseq |
Mouse Expression Set 430 |
1440839_x_at |
- |
- |
Mouse Expression Set 430 |
1440839_PM_x_at |
- |
- |
Murine Genome U74v2 Set |
166277_PM_at |
- |
- |
Murine Genome U74v2 Set |
166277_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1440839_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1440839_x_at |
- |
- |