Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929805_m1
          See other 1700099I09RIK GE Assays ›
          Gene Symbol:
          1700099I09Rik
          Gene Name:
          RIKEN cDNA 1700099I09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 102760255 - 102761896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929805_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700099I09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007100.1 - 1 - 2 32 71
          CN843001.1 - 1 - 2 43 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CD180 antigen Cd180 F630107B15,
          Ly78,
          RP105
          -53.6kb Sense Chr. 13 - 102693558 - 102706631 on NCBI Build GRCm38
          Downstream predicted gene 6211 Gm6211 EG621312 397.9kb Anti-sense Chr. 13 - 103159817 - 103160916 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2416762 AK007100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2416762 AV209598.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2416762 AK007100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2416762 AV209598.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10714958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11095290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29280756 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441817_PM_at - -
          Mouse Genome 430 2.0 Array 1441817_PM_at - -
          Mouse Genome 430 2.0 Array 1441817_at - -
          Mouse Expression Set 430 1441817_PM_at - -
          Mouse Expression Set 430 1441817_at - -
          Affymetrix HT Mouse Genome 430B Array 1441817_at - -
          Affymetrix HT Mouse Genome 430B Array 1441817_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline