Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700105F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007124.1 | - | 1 | 152 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | stress-associated endoplasmic reticulum protein family member 2 | Serp2 | 2810032E02Rik | -84.6kb | Sense | Chr. 14 - 76532812 - 76557919 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700108F19 gene | 1700108F19Rik |
1700020I02Rik, BB065030 |
34.7kb | Anti-sense | Chr. 14 - 76677568 - 76687108 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |