Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929924_m1
          See other 1810008I18RIK GE Assays ›
          Gene Symbol:
          1810008I18Rik
          Gene Name:
          RIKEN cDNA 1810008I18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 65804052 - 65806223 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929924_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810008I18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045301.1 1 - 2 280 77
          GenBank mRNA AK007376.1 - 1 - 2 281 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707476.1 - 1 - 2 276 Chr. 7 - 65804056 - 65806218 [+] on Build GRCm38 77
          GenBankmRNA AA289933.1 - 1 - 2 241 Chr. 7 - 65804090 - 65805959 [+] on Build GRCm38 77
          GenBankmRNA BY786390.1 - 1 - 2 299 Chr. 7 - 65804033 - 65805854 [+] on Build GRCm38 77
          GenBankmRNA BF582293.1 - 1 - 2 243 Chr. 7 - 65804089 - 65806234 [+] on Build GRCm38 77
          GenBankmRNA BE569317.1 - 1 - 2 291 Chr. 7 - 65804042 - 65806153 [+] on Build GRCm38 77
          GenBankmRNA BY776192.1 - 1 - 2 299 Chr. 7 - 65804033 - 65805870 [+] on Build GRCm38 77
          GenBankmRNA CB595507.1 - 2 - 3 346 Chr. 7 - 65803111 - 65806163 [+] on Build GRCm38 77
          GenBankmRNA BY146909.1 - 1 - 2 299 Chr. 7 - 65804033 - 65805859 [+] on Build GRCm38 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TM2 domain containing 3 Tm2d3 1110025I09Rik,
          5930422O05Rik,
          Blp2
          -102.1kb Sense Chr. 7 - 65693417 - 65701913 on NCBI Build GRCm38
          Downstream proprotein convertase subtilisin/kexin type 6 Pcsk6 C86343,
          Pace4,
          SPC4
          55.9kb Sense Chr. 7 - 65862136 - 66050386 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000987.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700009048.mm.1 TC0700000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009048.mm.1 NR_045301.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009048.mm.1 NR_045301.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009051.mm.1 TC0700000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009051.mm.1 NR_045301.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009051.mm.1 NR_045301.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009050.mm.1 TC0700000987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700009050.mm.1 NR_045301.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700009050.mm.1 NR_045301.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 874191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix