Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930181_g1
          See other 2010203P06RIK GE Assays ›
          Gene Symbol:
          2010203P06Rik
          Gene Name:
          RIKEN cDNA 2010203P06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 56595478 - 56601675 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930181_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010203P06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK008426.1 - 1 - 2 110 90
          AK008432.1 - 1 - 2 110 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY708335.1 - 1 - 2 110 Chr. 13 - 56595537 - 56596642 [+] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leukocyte cell-derived chemotaxin 2 Lect2 -47.0kb Anti-sense Chr. 13 - 56528959 - 56548561 on NCBI Build GRCm38
          Downstream transforming growth factor,
          beta induced
          Tgfbi 68kDa,
          AI181842,
          AI747162,
          Beta-ig,
          big-h3
          13.0kb Sense Chr. 13 - 56609603 - 56639339 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10639465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10829741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10505809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10827276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10914127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10829743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11083784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29062176 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 165928_PM_at - -
          Murine Genome U74v2 Set 165928_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 607015 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105860044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0