Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930226_s1
          See other 2210011G09RIK GE Assays ›
          Gene Symbol:
          2210011G09Rik
          Gene Name:
          RIKEN cDNA 2210011G09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930226_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2210011G09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK008706.1 - 1 548 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU526101.1 - 1 361 Chr. 2 - 80633929 - 80634470 [+] on Build GRCm38 89
          GenBankmRNA AV074931.1 - 1 119 Chr. 2 - 80634169 - 80634465 [+] on Build GRCm38 89
          GenBankmRNA BQ555490.1 - 1 Chr. 2 - 80633884 - 80634469 [-] on Build GRCm38 89
          GenBankmRNA BB777285.1 - 1 277 Chr. 2 - 80634011 - 80634465 [+] on Build GRCm38 89
          GenBankmRNA CA318748.1 - 1 174 Chr. 2 - 80634115 - 80634474 [+] on Build GRCm38 89
          GenBankmRNA BY708595.1 - 1 546 Chr. 2 - 80633740 - 80634440 [+] on Build GRCm38 89
          GenBankmRNA BY432907.1 - 1 230 Chr. 2 - 80634058 - 80634465 [+] on Build GRCm38 89
          GenBankmRNA BG970498.1 - 1 254 Chr. 2 - 80634034 - 80634809 [+] on Build GRCm38 89
          GenBankmRNA BY648891.1 - 1 262 Chr. 2 - 80634026 - 80634465 [+] on Build GRCm38 89
          GenBankmRNA BM115901.2 - 1 Chr. 2 - 80634158 - 80634469 [-] on Build GRCm38 89
          GenBankmRNA BB819196.1 - 1 289 Chr. 2 - 80634000 - 80634465 [+] on Build GRCm38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dual specificity phosphatase 19 Dusp19 5930436K22Rik,
          C79103,
          SKRP1,
          TS-DSP1
          -2.1kb Sense Chr. 2 - 80617214 - 80631661 on NCBI Build GRCm38
          Downstream nucleoporin 35 Nup35 2310006I24Rik,
          35kDa,
          5330402E05Rik,
          MP44,
          NO44
          4.3kb Sense Chr. 2 - 80638812 - 80660071 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P291304 AK008706.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 136584_PM_at - -
          Murine Genome U74v2 Set 136584_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 582142 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105220368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix