Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930545_m1
          See other 2310030A07RIK GE Assays ›
          Gene Symbol:
          2310030A07Rik
          Gene Name:
          RIKEN cDNA 2310030A07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 150320681 - 150346771 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930545_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310030A07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040603.1 2 - 3 186 73
          GenBank mRNA AK009534.1 - 2 - 3 187 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY709227.1 - 2 - 3 187 Chr. 1 - 150320681 - 150346771 [-] on Build GRCm38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream prostaglandin-endoperoxide synthase 2,
          opposite strand 2
          Ptgs2os2 Gm26687,
          Linc-cox2,
          Lincrna-cox2
          -157.7kb Sense Chr. 1 - 150159043 - 150164948 on NCBI Build GRCm38
          Downstream cDNA sequence BC003331 BC003331 1810053E15Rik,
          Odr4,
          odr-4
          16.3kb Sense Chr. 1 - 150361311 - 150393055 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003238.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45595 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04303646 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04303637 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131973 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04303646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20708283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100026185.mm.1 TC0100003238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100026185.mm.1 NR_040603.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026185.mm.1 NR_040603.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026186.mm.1 TC0100003238.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100026186.mm.1 NR_040603.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100026186.mm.1 NR_040603.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 520946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0