Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930547_m1
          See other 2310031A07RIK GE Assays ›
          Gene Symbol:
          2310031A07Rik
          Gene Name:
          RIKEN cDNA 2310031A07 gene
          Gene Aliases:
          AV087459
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930547_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310031A07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK009549.1 - 1 - 2 200 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY709238.1 - 1 - 2 200 Chr. 11 - 46440450 - 46447265 [-] on Build GRCm38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mediator complex subunit 7 Med7 1110063B05Rik,
          Crsp33,
          Crsp9
          Anti-sense Chr. 11 - 46436933 - 46442721 on NCBI Build GRCm38
          Downstream hepatitis A virus cellular receptor 2 Havcr2 TIM-3,
          Tim3,
          Timd3
          7.3kb Anti-sense Chr. 11 - 46454547 - 46481255 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127994 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139478 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133015 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133015 - -
          Whole Mouse Genome G4122A A_52_P453411 AK009549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453411 CK030901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453411 CK788351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453411 BC011333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453411 BE333513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P649590 AK088386.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133015 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430419_PM_at - -
          Mouse Genome 430 2.0 Array 1430419_PM_at - -
          Mouse Genome 430 2.0 Array 1430419_at - -
          Mouse Expression Set 430 1430419_at - -
          Mouse Expression Set 430 1430419_PM_at - -
          Murine Genome U74v2 Set 167036_PM_at - -
          Murine Genome U74v2 Set 167036_at - -
          Mouse Genome 430A 2.0 Array 1430419_PM_at - -
          Mouse Genome 430A 2.0 Array 1430419_at - -
          Affymetrix HT Mouse Genome 430A Array 1430419_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1430419_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline