Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930711_s1
          See other A930024O17RIK GE Assays ›
          Gene Symbol:
          A930024O17Rik
          Gene Name:
          RIKEN cDNA A930024O17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930711_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930024O17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020888.1 - 1 753 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU502958.1 - 1 299 Chr. 11 - 117385046 - 117385393 [+] on Build GRCm38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream septin 9 Sept9 MSF1,
          Msf,
          PNUTL4,
          Sint1
          -22.2kb Sense Chr. 11 - 117199661 - 117362325 on NCBI Build GRCm38
          Downstream predicted gene 11732 Gm11732 OTTMUSG00000003800 40.8kb Sense Chr. 11 - 117426163 - 117438866 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09488687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09643281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09170688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09565292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28164895 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 648940 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105290373 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0