Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930886_s1
          See other B230114A03RIK GE Assays ›
          Gene Symbol:
          B230114A03Rik
          Gene Name:
          RIKEN cDNA B230114A03 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930886_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230114A03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020976.1 - 1 1148 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          29907
          Gm29907 Sense Chr. 16 - 39984595 - 40264481 on NCBI Build GRCm38
          Downstream limbic system-associated membrane protein Lsamp 5430428I19,
          AW046674,
          B130007O04Rik,
          D930023J12Rik,
          Lam,
          Lamp
          1.4mb Sense Chr. 16 - 41532788 - 42181679 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12793781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12793763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30694581 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433200_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433199_PM_at - -
          Mouse Genome 430 2.0 Array 1433200_PM_at - -
          Mouse Genome 430 2.0 Array 1433199_PM_at - -
          Mouse Genome 430 2.0 Array 1433199_at - -
          Mouse Genome 430 2.0 Array 1433200_at - -
          Mouse Expression Set 430 1433199_PM_at - -
          Mouse Expression Set 430 1433199_at - -
          Mouse Expression Set 430 1433200_PM_at - -
          Mouse Expression Set 430 1433200_at - -
          Affymetrix HT Mouse Genome 430B Array 1433199_at - -
          Affymetrix HT Mouse Genome 430B Array 1433200_at - -
          Affymetrix HT Mouse Genome 430B Array 1433200_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433199_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104540286 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0