Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930966_s1
          See other CD47 GE Assays ›
          Gene Symbol:
          Cd47
          Gene Name:
          CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
          Gene Aliases:
          9130415E20Rik, AA407862, AI848868, AW108519, B430305P08Rik, IAP, Itgp
          Chromosome Location:
          Chr.16: 49855252 - 49912424 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03930966_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd47
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021021.1 - 1 653 63
          AK164165.1 - 671 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE687287.1 - 1 370 Chr. 16 - 49911745 - 49912211 [+] on Build GRCm38 63
          GenBankmRNA BE631052.1 - 1 316 Chr. 16 - 49911878 - 49912206 [+] on Build GRCm38 63
          GenBankmRNA BY181641.1 - 1 232 Chr. 16 - 49911883 - 49912215 [+] on Build GRCm38 63
          GenBankmRNA BY376973.1 - 1 220 Chr. 16 - 49911897 - 49912244 [+] on Build GRCm38 63
          GenBankmRNA BE852404.1 - 1 Chr. 16 - 49912049 - 49912252 [-] on Build GRCm38 63
          GenBankmRNA BQ550511.1 - 1 483 Chr. 16 - 49911633 - 49912252 [+] on Build GRCm38 63
          GenBankmRNA BP754330.1 - 1 326 Chr. 16 - 49911814 - 49912577 [+] on Build GRCm38 63
          GenBankmRNA BE992312.1 - 1 Chr. 16 - 49911968 - 49912268 [-] on Build GRCm38 63
          GenBankmRNA AA177747.1 - 1 108 Chr. 16 - 49912007 - 49912455 [+] on Build GRCm38 63
          GenBankmRNA BQ550510.1 - 1 Chr. 16 - 49911961 - 49912252 [-] on Build GRCm38 63
          GenBankmRNA BU702662.1 - 1 656 Chr. 16 - 49911506 - 49912194 [+] on Build GRCm38 63
          GenBankmRNA BB661184.1 - 1 232 Chr. 16 - 49911883 - 49912530 [+] on Build GRCm38 63
          GenBankmRNA AA071831.1 - 1 292 Chr. 16 - 49911822 - 49912252 [+] on Build GRCm38 63
          GenBankmRNA CF553966.1 - 1 334 Chr. 16 - 49911788 - 49912268 [+] on Build GRCm38 63
          GenBankmRNA BI250660.1 - 1 101 Chr. 16 - 49912026 - 49912264 [+] on Build GRCm38 63
          GenBankmRNA BE632825.1 - 1 Chr. 16 - 49911980 - 49912252 [-] on Build GRCm38 63
          GenBankmRNA CF539264.1 - 1 522 Chr. 16 - 49911593 - 49912268 [+] on Build GRCm38 63
          GenBankmRNA BM937913.1 - 1 42 Chr. 16 - 49912073 - 49912759 [+] on Build GRCm38 63
          GenBankmRNA BE850588.1 - 1 98 Chr. 16 - 49912017 - 49912235 [+] on Build GRCm38 63
          GenBankmRNA BE848646.1 - 1 381 Chr. 16 - 49911734 - 49912190 [+] on Build GRCm38 63
          GenBankmRNA BF454475.1 - 1 352 Chr. 16 - 49911748 - 49912187 [+] on Build GRCm38 63
          GenBankmRNA BE992334.1 - 1 Chr. 16 - 49911951 - 49912268 [-] on Build GRCm38 63
          GenBankmRNA CJ180215.1 - 1 231 Chr. 16 - 49911884 - 49912252 [+] on Build GRCm38 63
          GenBankmRNA CJ136935.1 - 1 231 Chr. 16 - 49911884 - 49912569 [+] on Build GRCm38 63
          GenBankmRNA BY177317.1 - 1 93 Chr. 16 - 49912022 - 49912362 [+] on Build GRCm38 63
          GenBankmRNA BB864279.1 - 1 236 Chr. 16 - 49911881 - 49912314 [+] on Build GRCm38 63
          GenBankmRNA CB843524.2 - 235 Chr. - - [-] on Build 63
          GenBankmRNA BU056052.1 - 1 388 Chr. 16 - 49911727 - 49912268 [+] on Build GRCm38 63
          GenBankmRNA CB723661.1 - 1 445 Chr. 16 - 49911670 - 49912268 [+] on Build GRCm38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 4633 Gm4633 -56.4kb Anti-sense Chr. 16 - 49811731 - 49855046 on NCBI Build GRCm38
          Downstream predicted gene 5486 Gm5486 EG433024 17.2kb Anti-sense Chr. 16 - 49929454 - 49972476 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 435912 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0