Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03931199_s1
          See other C030039E19RIK GE Assays ›
          Gene Symbol:
          C030039E19Rik
          Gene Name:
          RIKEN cDNA C030039E19 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03931199_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C030039E19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK021134.1 - 1 550 109
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY607232.1 - 1 263 Chr. 13 - 29015743 - 29016122 [+] on Build GRCm38 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SRY (sex determining region Y)-box 4 Sox4 AA682046,
          Sox-4
          -61.8kb Anti-sense Chr. 13 - 28950716 - 28953682 on NCBI Build GRCm38
          Downstream CDK5 regulatory subunit associated protein 1-like 1 Cdkal1 1190005B03Rik,
          6620401C13Rik
          175.6kb Anti-sense Chr. 13 - 29191751 - 29855684 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708712 - -
          Whole Mouse Genome G4122A A_51_P216029 AK021134.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10565277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10972500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10565275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10649625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10806631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10649610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28909228 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106860338 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0