Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03932243_s1
          See other BC020402 GE Assays ›
          Gene Symbol:
          BC020402
          Gene Name:
          cDNA sequence BC020402
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 7678971 - 7681158 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03932243_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC020402
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033219.1 1 1071 74
          GenBank mRNA AK030763.1 - 1 954 74
          AK132475.1 - 1 1072 74
          BC020402.1 - 1 1038 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU925196.1 - 1 200 Chr. 10 - 7679684 - 7680291 [-] on Build GRCm38 74
          GenBankmRNA BB029175.2 - 1 521 Chr. 10 - 7679941 - 7680589 [-] on Build GRCm38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein-L-isoaspartate (D-aspartate) O-methyltransferase 1 Pcmt1 C79501,
          PIMT
          Sense Chr. 10 - 7629380 - 7680973 on NCBI Build GRCm38
          Downstream large tumor suppressor Lats1 AW208599 168.0 Anti-sense Chr. 10 - 7681209 - 7716461 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001698.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000012426.mm.1 TC1000001698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012426.mm.1 NR_033219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012426.mm.1 NR_033219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012425.mm.1 TC1000001698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012425.mm.1 NR_033219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012425.mm.1 NR_033219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012424.mm.1 TC1000001698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012424.mm.1 NR_033219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012424.mm.1 NR_033219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012422.mm.1 TC1000001698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012422.mm.1 NR_033219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012422.mm.1 NR_033219.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012423.mm.1 TC1000001698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000012423.mm.1 NR_033219.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000012423.mm.1 NR_033219.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0