Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03932448_s1
          See other A730054J21RIK GE Assays ›
          Gene Symbol:
          A730054J21Rik
          Gene Name:
          RIKEN cDNA A730054J21 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03932448_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730054J21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK031124.1 - 1 1656 102
          AW045500.1 - 1 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM118773.2 - 1 Chr. 1 - 178938841 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BB695903.1 - 1 484 Chr. 1 - 178938675 - 178939199 [+] on Build GRCm38 102
          GenBankmRNA BM119403.2 - 1 Chr. 1 - 178938890 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BM119292.2 - 1 Chr. 1 - 178938568 - 178939200 [-] on Build GRCm38 102
          GenBankmRNA BM119567.2 - 1 Chr. 1 - 178938572 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BM231497.2 - 1 Chr. 1 - 178938650 - 178939199 [-] on Build GRCm38 101
          GenBankmRNA BM119295.2 - 1 Chr. 1 - 178938568 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BQ557013.1 - 1 Chr. 1 - 178938544 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BM118542.1 - 1 Chr. 1 - 178938747 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BM231081.2 - 1 Chr. 1 - 178938615 - 178939199 [-] on Build GRCm38 101
          GenBankmRNA BM119126.2 - 1 Chr. 1 - 178938638 - 178939199 [-] on Build GRCm38 102
          GenBankmRNA BB035949.2 - 1 657 Chr. 1 - 178938504 - 178939199 [+] on Build GRCm38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream kinesin family member 26B Kif26b 4832420M10,
          BC056349,
          D230039L06Rik
          Sense Chr. 1 - 178525741 - 178939200 on NCBI Build GRCm38
          Downstream SET and MYND domain containing 3 Smyd3 2410008A19Rik,
          Zmynd1
          12.8kb Anti-sense Chr. 1 - 178951960 - 179518052 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82055 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122895 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122895 - -
          Whole Mouse Genome G4122A A_51_P103586 AK031124.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103586 AK043086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103586 BC023483.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100340 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20867023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20867020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20867024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00727607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20867025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20867019 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1438531_PM_at - -
          Mouse Genome 430 2.0 Array 1459304_PM_at - -
          Mouse Genome 430 2.0 Array 1438531_at - -
          Mouse Genome 430 2.0 Array 1459304_at - -
          Mouse Genome 430 2.0 Array 1438531_PM_at - -
          Mouse Expression Set 430 1438531_at - -
          Mouse Expression Set 430 1459304_PM_at - -
          Mouse Expression Set 430 1438531_PM_at - -
          Mouse Expression Set 430 1459304_at - -
          Murine Genome U74v2 Set 137699_at - -
          Murine Genome U74v2 Set 137699_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438531_at - -
          Affymetrix HT Mouse Genome 430B Array 1438531_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix