Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03932538_s1
          See other 6030443J06RIK GE Assays ›
          Gene Symbol:
          6030443J06Rik
          Gene Name:
          RIKEN cDNA 6030443J06 gene
          Gene Aliases:
          1700030G06Rik
          Chromosome Location:
          Chr.5: 22550413 - 22558171 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03932538_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6030443J06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102315.1 3 3315 68
          GenBank mRNA AK029857.1 - 3 3292 68
          AK031505.1 - 1 3248 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB039591.2 - 1 481 Chr. 5 - 22556682 - 22557363 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream origin recognition complex,
          subunit 5
          Orc5 AL033327,
          MmORC5,
          Orc5l
          -3.5kb Anti-sense Chr. 5 - 22486484 - 22550367 on NCBI Build GRCm38
          Downstream RIKEN cDNA A930003O13 gene A930003O13Rik 181.5kb Anti-sense Chr. 5 - 22738910 - 22746884 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000186.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2282984 AK029857.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760427 D89901.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580194 AK009246.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580194 NM_001081226.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47676 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150086 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126496 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110350 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156152 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2282984 AK029857.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05503110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05503097 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502769 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01052995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502998 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01053133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502935 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01053092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502831 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05503019 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110350 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502720 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01053008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502745 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01053210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05502771 - -
          Whole Mouse Genome G4122A A_52_P38128 AK030448.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P38128 AK006545.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P38128 AK044020.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107608 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05502831 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01053008 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01053133 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126496 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05502745 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156152 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04599672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04128078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04202268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04153634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04239082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04301830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04301829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04239092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04153632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04714305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04252078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04202254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04440185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04865359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04153625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04687941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23532333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04740540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500001333.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001333.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001338.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001338.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001341.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001341.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001335.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001335.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001339.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001339.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001340.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001340.mm.1 NR_102315.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001336.mm.1 TC0500000186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001336.mm.1 NR_102315.1 Refseq
          Murine Genome U74v2 Set 134523_at - -
          Murine Genome U74v2 Set 134523_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 392093 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0