Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03932971_s1
          See other A930011O12RIK GE Assays ›
          Gene Symbol:
          A930011O12Rik
          Gene Name:
          RIKEN cDNA A930011O12 gene
          Gene Aliases:
          Rncr3, mir-124-1, mir-3078
          Chromosome Location:
          Chr.14: 64588115 - 64593961 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03932971_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930011O12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040709.1 4 3808 62
          GenBank mRNA AK032410.1 - 1 1445 62
          BC030402.1 - 2 3593 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ921767.1 - 1 296 Chr. 14 - 64593435 - 64593957 [+] on Build GRCm38 62
          GenBankmRNA BU529846.1 - 1 152 Chr. 14 - 64593581 - 64593966 [+] on Build GRCm38 62
          GenBankmRNA BG298685.1 - 1 469 Chr. 14 - 64593266 - 64593909 [+] on Build GRCm38 62
          GenBankmRNA BE949994.1 - 1 Chr. 14 - 64593699 - 64593963 [-] on Build GRCm38 62
          GenBankmRNA BI730001.1 - 114 Chr. - - [-] on Build 62
          GenBankmRNA BQ178251.1 - 1 472 Chr. 14 - 64593258 - 64593928 [+] on Build GRCm38 62
          GenBankmRNA BB677946.1 - 1 201 Chr. 14 - 64593530 - 64593959 [+] on Build GRCm38 62
          GenBankmRNA EL963240.1 - 1 Chr. 14 - 64593446 - 64593963 [-] on Build GRCm38 62
          GenBankmRNA BF457332.1 - 1 Chr. 14 - 64593602 - 64593929 [-] on Build GRCm38 62
          GenBankmRNA CD774275.1 - 1 Chr. 14 - 64593257 - 64593963 [-] on Build GRCm38 62
          GenBankmRNA BB800194.1 - 1 171 Chr. 14 - 64593560 - 64593959 [+] on Build GRCm38 62
          GenBankmRNA CD741372.1 - 1 Chr. 14 - 64593281 - 64593963 [-] on Build GRCm38 62
          GenBankmRNA CO042523.1 - 1 Chr. 14 - 64593254 - 64593955 [-] on Build GRCm38 62
          GenBankmRNA BI872857.1 - 1 246 Chr. 14 - 64593485 - 64593963 [+] on Build GRCm38 62
          GenBankmRNA AV343709.2 - 1 443 Chr. 14 - 64593295 - 64593959 [+] on Build GRCm38 62
          GenBankmRNA BY629680.1 - 1 156 Chr. 14 - 64593575 - 64593962 [+] on Build GRCm38 62
          GenBankmRNA BG295392.1 - 1 433 Chr. 14 - 64593369 - 64593957 [+] on Build GRCm38 62
          GenBankmRNA BB804539.1 - 1 147 Chr. 14 - 64593584 - 64593959 [+] on Build GRCm38 62
          GenBankmRNA BU505303.1 - 1 625 Chr. 14 - 64593106 - 64593958 [+] on Build GRCm38 62
          GenBankmRNA BY648920.1 - 1 219 Chr. 14 - 64593512 - 64593959 [+] on Build GRCm38 62
          GenBankmRNA BI738316.1 - 1 115 Chr. 14 - 64593615 - 64593963 [+] on Build GRCm38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 3078 Mir3078 mmu-mir-3078 -1.0kb Sense Chr. 14 - 64591185 - 64591271 on NCBI Build GRCm38
          Downstream kinesin family member 13B Kif13b 5330429L19Rik,
          6030414C01,
          AI429803,
          C130021D12Rik,
          GAKIN
          58.5kb Sense Chr. 14 - 64652490 - 64809618 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000923.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02869089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02869082 - -
          Whole Mouse Genome G4122A A_51_P364121 BC030402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P364121 AK044422.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106262 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02869082 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435453_PM_at - -
          Mouse Genome 430 2.0 Array 1435453_PM_at - -
          Mouse Genome 430 2.0 Array 1435453_at - -
          Affymetrix Mouse Clariom S Array PSR1400007353.mm.1 TC1400000923.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400007353.mm.1 NR_040709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400007353.mm.1 NR_040709.1 Refseq
          Mouse Expression Set 430 1435453_PM_at - -
          Mouse Expression Set 430 1435453_at - -
          Murine Genome U74v2 Set 133118_at - -
          Murine Genome U74v2 Set 138139_at - -
          Murine Genome U74v2 Set 133118_PM_at - -
          Murine Genome U74v2 Set 138139_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435453_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435453_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 839053 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0