Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03933244_s1
          See other FAM101B GE Assays ›
          Gene Symbol:
          Fam101b
          Gene Name:
          family with sequence similarity 101, member B
          Gene Aliases:
          1500005K14Rik, RefilinB, cfm
          Chromosome Location:
          Chr.11: 76019194 - 76027782 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03933244_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam101b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029658.1 NP_083934.1 3 3203 69
          GenBank mRNA AK033150.1 - 1 2453 69
          W81812.1 - 1 93 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA795108.1 - 1 101 Chr. 11 - 76019202 - 76019603 [-] on Build GRCm38 69
          GenBankmRNA AA727879.1 - 1 246 Chr. 11 - 76019268 - 76019750 [-] on Build GRCm38 69
          GenBankmRNA BB306438.2 - 1 469 Chr. 11 - 76019291 - 76019970 [-] on Build GRCm38 69
          GenBankmRNA BB065981.2 - 1 355 Chr. 11 - 76019203 - 76019856 [-] on Build GRCm38 69
          GenBankmRNA AU021661.1 - 1 Chr. 11 - 76019291 - 76019696 [+] on Build GRCm38 69
          GenBankmRNA BX636985.1 - 1 Chr. 11 - 76019246 - 76019624 [+] on Build GRCm38 69
          GenBankmRNA BU700780.1 - 1 Chr. 11 - 76019194 - 76019791 [+] on Build GRCm38 69
          GenBankmRNA BE691555.1 - 1 Chr. 11 - 76019197 - 76019564 [+] on Build GRCm38 69
          GenBankmRNA AI430485.1 - 1 390 Chr. 11 - 76019388 - 76019892 [-] on Build GRCm38 69
          GenBankmRNA AA572082.1 - 1 343 Chr. 11 - 76019462 - 76019847 [-] on Build GRCm38 69
          GenBankmRNA W62938.1 - 541 Chr. - - [-] on Build 69
          GenBankmRNA DV060887.1 - 396 Chr. - - [-] on Build 69
          GenBankmRNA AW744720.1 - 1 Chr. 11 - 76019194 - 76019623 [+] on Build GRCm38 69
          GenBankmRNA AA416121.1 - 1 229 Chr. 11 - 76019369 - 76019732 [-] on Build GRCm38 69
          GenBankmRNA AA879802.1 - 1 253 Chr. 11 - 76019377 - 76019755 [-] on Build GRCm38 69
          GenBankmRNA CO040051.1 - 1 Chr. 11 - 76019194 - 76019866 [+] on Build GRCm38 69
          GenBankmRNA BG796024.1 - 1 Chr. 11 - 76019379 - 76019825 [+] on Build GRCm38 69
          GenBankmRNA AA145478.1 - 1 408 Chr. 11 - 76019436 - 76019910 [-] on Build GRCm38 69
          GenBankmRNA AA856446.1 - 1 487 Chr. 11 - 76019467 - 76019989 [-] on Build GRCm38 69
          GenBankmRNA AI197589.1 - 1 228 Chr. 11 - 76019350 - 76019732 [-] on Build GRCm38 69
          GenBankmRNA AI413094.1 - 1 Chr. 11 - 76019194 - 76019555 [+] on Build GRCm38 69
          GenBankmRNA BE691576.1 - 1 Chr. 11 - 76019219 - 76019564 [+] on Build GRCm38 69
          GenBankmRNA BG795014.1 - 1 Chr. 11 - 76019379 - 76019831 [+] on Build GRCm38 69
          GenBankmRNA AA880424.1 - 1 246 Chr. 11 - 76019376 - 76019749 [-] on Build GRCm38 69
          GenBankmRNA BE634108.1 - 1 95 Chr. 11 - 76019214 - 76019597 [-] on Build GRCm38 69
          GenBankmRNA AA684526.1 - 1 93 Chr. 11 - 76019192 - 76019596 [-] on Build GRCm38 69
          GenBankmRNA AI646280.1 - 1 Chr. 11 - 76019291 - 76019667 [+] on Build GRCm38 69
          GenBankmRNA W48101.1 - 437 Chr. - - [-] on Build 69
          GenBankmRNA AA423223.1 - 1 187 Chr. 11 - 76019269 - 76019689 [-] on Build GRCm38 69
          GenBankmRNA CJ319524.1 - 1 248 Chr. 11 - 76019292 - 76019750 [-] on Build GRCm38 69
          GenBankmRNA W81907.1 - 389 Chr. - - [-] on Build 69
          GenBankmRNA AA041611.1 - 1 389 Chr. 11 - 76019453 - 76019892 [-] on Build GRCm38 69
          GenBankmRNA BF457768.1 - 1 Chr. 11 - 76019280 - 76019709 [+] on Build GRCm38 69
          GenBankmRNA BG070087.2 - 1 Chr. 11 - 76019429 - 76019800 [+] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700016K19 gene 1700016K19Rik Liat1 -15.6kb Anti-sense Chr. 11 - 75999912 - 76003569 on NCBI Build GRCm38
          Downstream vacuolar protein sorting 53 (yeast) Vps53 2010002A08Rik,
          2310040I21Rik,
          3100002B05Rik,
          Hccs1
          24.3kb Sense Chr. 11 - 76046226 - 76179630 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial to mesenchymal transition
          actin cytoskeleton organization
          skeletal system morphogenesis
          negative regulation of chondrocyte development
          actin filament bundle organization
          negative regulation of bone mineralization involved in bone maturation
          filamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK005164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK033150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK171075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 NM_029658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK005164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK033150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 AK171075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P275697 NM_029658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P5891 AK033150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P5891 NM_029658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P5891 AK033150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P5891 NM_029658.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105131 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167179 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P5891 AK033150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P5891 NM_029658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P5891 AK033150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P5891 NM_029658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK005164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK033150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK171075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 NM_029658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK005164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK033150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 AK171075.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P275697 NM_029658.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02366145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167179 - -
          Whole Mouse Genome G4122A A_51_P275697 AK005164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 AK033150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 AK171075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 NM_029658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 AK005164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 AK033150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 AK171075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P275697 NM_029658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P5891 AK033150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P5891 NM_029658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P5891 AK033150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P5891 NM_029658.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09497934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09311998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09158516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09374668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09522398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09522387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09415534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09323160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09158517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09497933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09158511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09647246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09522394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09189505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27929555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09533305 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429765_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456603_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429764_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388488 - -
          Mouse Genome 430 2.0 Array 1456603_at - -
          Mouse Genome 430 2.0 Array 1429765_PM_at - -
          Mouse Genome 430 2.0 Array 1456603_PM_at - -
          Mouse Genome 430 2.0 Array 1429765_at - -
          Mouse Genome 430 2.0 Array 1429764_at - -
          Mouse Genome 430 2.0 Array 1429764_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76566_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76566_PM_at - -
          Mouse Expression Set 430 1456603_PM_at - -
          Mouse Expression Set 430 1456603_at - -
          Mouse Expression Set 430 1429765_at - -
          Mouse Expression Set 430 1429765_PM_at - -
          Mouse Expression Set 430 1429764_at - -
          Mouse Expression Set 430 1429764_PM_at - -
          Murine Genome U74v2 Set 96791_PM_at - -
          Murine Genome U74v2 Set 96791_at - -
          Mouse Genome 430A 2.0 Array 1456603_PM_at - -
          Mouse Genome 430A 2.0 Array 1456603_at - -
          Affymetrix HT Mouse Genome 430A Array 1456603_at - -
          Affymetrix HT Mouse Genome 430A Array 1456603_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429765_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429765_at - -
          Affymetrix HT Mouse Genome 430B Array 1429764_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 747352 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2490729 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217606 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0