Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03934654_s1
          See other RNF165 GE Assays ›
          Gene Symbol:
          Rnf165
          Gene Name:
          ring finger protein 165
          Gene Aliases:
          2900024M11Rik, AI427432, Akd2, Ark2c, G630064H08Rik, Gm96
          Chromosome Location:
          Chr.18: 77456110 - 77566378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03934654_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf165
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164504.1 NP_001157976.1 8 4079 97
          GenBank mRNA AK036050.1 - 1 953 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF746146.1 - 1 596 Chr. 18 - 77459178 - 77459915 [-] on Build GRCm38 97
          GenBankmRNA BI688802.1 - 1 483 Chr. 18 - 77459040 - 77459813 [-] on Build GRCm38 97
          GenBankmRNA BG800205.1 - 1 551 Chr. 18 - 77459283 - 77459880 [-] on Build GRCm38 97
          GenBankmRNA CB247980.1 - 1 555 Chr. 18 - 77459130 - 77459884 [-] on Build GRCm38 97
          GenBankmRNA AA967881.1 - 1 270 Chr. 18 - 77459235 - 77459600 [-] on Build GRCm38 97
          GenBankmRNA BX528152.1 - 1 271 Chr. 18 - 77459076 - 77459600 [-] on Build GRCm38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream lipoxygenase homology domains 1 Loxhd1 1700096C21Rik,
          4932417K07,
          sba
          -15.4kb Anti-sense Chr. 18 - 77281949 - 77442932 on NCBI Build GRCm38
          Downstream RIKEN cDNA 8030462N17 gene 8030462N17Rik Akd2,
          Ark2N
          171.2kb Sense Chr. 18 - 77631488 - 77714489 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001625.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          axonogenesis
          motor neuron axon guidance
          multicellular organism aging
          protein catabolic process
          positive regulation of BMP signaling pathway
          forelimb morphogenesis
          limb development
          innervation
          muscle structure development
          protein binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800011693.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011693.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011693.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011692.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011692.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011692.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011691.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011691.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011691.mm.1 NM_001164504.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0