Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03934768_s1
          See other ZC3HAV1 GE Assays ›
          Gene Symbol:
          Zc3hav1
          Gene Name:
          zinc finger CCCH type, antiviral 1
          Gene Aliases:
          1200014N16Rik, 2900058M19Rik, 9130009D18Rik, 9830115L13Rik, ARTD13, D6Bwg1452e, ZAP
          Chromosome Location:
          Chr.6: 38305286 - 38355840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03934768_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zc3hav1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK036481.1 - 1 1755 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA636972.1 - 1 54 Chr. 6 - 38305286 - 38305456 [-] on Build GRCm38 96
          GenBankmRNA BY589165.1 - 1 345 Chr. 6 - 38305301 - 38305747 [-] on Build GRCm38 96
          GenBankmRNA BE689448.1 - 1 309 Chr. 6 - 38305300 - 38305711 [-] on Build GRCm38 96
          GenBankmRNA AI646027.1 - 1 Chr. 6 - 38305300 - 38305748 [+] on Build GRCm38 96
          GenBankmRNA BE137294.1 - 1 309 Chr. 6 - 38305317 - 38305711 [-] on Build GRCm38 96
          GenBankmRNA BE200241.1 - 1 Chr. 6 - 38305300 - 38305617 [+] on Build GRCm38 96
          GenBankmRNA BB757349.1 - 1 434 Chr. 6 - 38305297 - 38305834 [-] on Build GRCm38 95
          GenBankmRNA BE198885.1 - 1 309 Chr. 6 - 38305300 - 38305711 [-] on Build GRCm38 96
          GenBankmRNA BB134490.2 - 1 572 Chr. 6 - 38305323 - 38305976 [-] on Build GRCm38 96
          GenBankmRNA BB761715.1 - 1 342 Chr. 6 - 38305301 - 38305741 [-] on Build GRCm38 96
          GenBankmRNA CB845072.2 - 1 494 Chr. 6 - 38305305 - 38305885 [-] on Build GRCm38 96
          GenBankmRNA BF120231.1 - 1 78 Chr. 6 - 38305306 - 38305474 [-] on Build GRCm38 96
          GenBankmRNA BG092916.1 - 1 Chr. 6 - 38305300 - 38305569 [+] on Build GRCm38 96
          GenBankmRNA BI665593.1 - 1 168 Chr. 6 - 38273254 - 38305570 [-] on Build GRCm38 96
          GenBankmRNA BG230228.1 - 1 Chr. 6 - 38305286 - 38305627 [+] on Build GRCm38 96
          GenBankmRNA BX517258.1 - 1 310 Chr. 6 - 38305300 - 38305711 [-] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger CCCH-type,
          antiviral 1-like
          Zc3hav1l B130055L09Rik,
          E430016P22Rik
          -6.0kb Sense Chr. 6 - 38287394 - 38299259 on NCBI Build GRCm38
          Downstream tetratricopeptide repeat domain 26 Ttc26 9330141E21Rik,
          9430097H08Rik
          74.3kb Anti-sense Chr. 6 - 38381474 - 38427647 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0