Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03935113_s1
          See other 6030442K20RIK GE Assays ›
          Gene Symbol:
          6030442K20Rik
          Gene Name:
          RIKEN cDNA 6030442K20 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03935113_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6030442K20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK037610.1 - 1 1450 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dual serine/threonine and tyrosine protein kinase Dstyk A930019K20Rik,
          C430014H23Rik,
          C820013G01,
          Ripk5
          -8.4kb Anti-sense Chr. 1 - 132417181 - 132466959 on NCBI Build GRCm38
          Downstream retinoblastoma binding protein 5 Rbbp5 4933411J24Rik,
          C330016J05
          322.0 Anti-sense Chr. 1 - 132477347 - 132505665 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136605 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141733 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141733 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109154 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432866_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454391_PM_at - -
          Mouse Genome 430 2.0 Array 1454391_at - -
          Mouse Genome 430 2.0 Array 1454391_PM_at - -
          Mouse Genome 430 2.0 Array 1432866_PM_at - -
          Mouse Genome 430 2.0 Array 1432866_at - -
          Mouse Expression Set 430 1454391_at - -
          Mouse Expression Set 430 1432866_at - -
          Mouse Expression Set 430 1432866_PM_at - -
          Mouse Expression Set 430 1454391_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432866_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432866_at - -
          Affymetrix HT Mouse Genome 430B Array 1454391_at - -
          Affymetrix HT Mouse Genome 430B Array 1454391_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106660279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix