Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03935719_s1
          See other B630006K09RIK GE Assays ›
          Gene Symbol:
          B630006K09Rik
          Gene Name:
          RIKEN cDNA B630006K09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03935719_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B630006K09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK037902.1 - 1 1791 92
          AK039810.1 - 1 348 92
          AK046747.1 - 1 1976 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB634214.1 - 1 348 Chr. 6 - 41568922 - 41569565 [+] on Build GRCm38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          38582
          Gm38582 Sense Chr. 6 - 41558555 - 41603450 on NCBI Build GRCm38
          Downstream Eph receptor B6 Ephb6 Cekl,
          Mep
          35.0kb Sense Chr. 6 - 41605040 - 41620507 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795238 AK038977.1 genbank_mrna_mouse
          Whole Mouse Genome G4122A A_52_P660722 AK037902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660722 AK039810.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660722 AK046747.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05276785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05205779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05078561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05846704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05454503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05078562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24394453 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821080 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102320504 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline