Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936057_m1
          See other GM4668 GE Assays ›
          Gene Symbol:
          Gm4668
          Gene Name:
          predicted gene 4668
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 116184530 - 116195049 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936057_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4668
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK041339.1 - 2 - 3 435 54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB227220.2 - 1 - 2 192 Chr. 9 - 116190822 - 116194870 [+] on Build GRCm38 54

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transforming growth factor,
          beta receptor II
          Tgfbr2 1110020H15Rik,
          AU042018,
          DNIIR,
          RIIDN,
          TBR-II,
          TbetaR-II,
          TbetaRII
          -15.4kb Anti-sense Chr. 9 - 116087695 - 116175363 on NCBI Build GRCm38
          Downstream RIKEN cDNA D730003K21 gene D730003K21Rik 134.3kb Sense Chr. 9 - 116327308 - 116339659 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P714949 AK041339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P714949 AK041339.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449688 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P714949 AK041339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P714949 AK041339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P714949 AK041339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P714949 AK041339.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07608608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07544264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07545346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07656340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08189531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07642587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07512660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07666012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08189530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07863131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07642588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07946352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07544265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07656343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26853784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5756d5bff6-86w5v:80/100.66.130.62:80.
          git-commit: 27702359083d4c7b0962b032f2611a59cac079b5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0