Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936378_g1
          See other A730043L09RIK GE Assays ›
          Gene Symbol:
          A730043L09Rik
          Gene Name:
          RIKEN cDNA A730043L09 gene
          Gene Aliases:
          A730043L09
          Chromosome Location:
          Chr.9: 62242596 - 62244100 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936378_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730043L09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040769.1 1 - 2 156 168
          GenBank mRNA AK042959.1 - 1 - 2 157 168

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glucuronyl C5-epimerase Glce 1110017N23Rik,
          AI747411,
          C130034A12Rik,
          Hsepi
          -121.0kb Sense Chr. 9 - 62057248 - 62122720 on NCBI Build GRCm38
          Downstream sperm equatorial segment protein 1 Spesp1 4921508E09Rik,
          AI428149
          26.6kb Sense Chr. 9 - 62270729 - 62282179 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002458.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350337 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07803578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07967249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07983621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07749369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07803577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07967252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07628859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08098371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07749367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07803583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26569521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900020581.mm.1 TC0900002458.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020581.mm.1 NR_040769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020581.mm.1 NR_040769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020584.mm.1 TC0900002458.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020584.mm.1 NR_040769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020584.mm.1 NR_040769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020576.mm.1 TC0900002458.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020576.mm.1 NR_040769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020576.mm.1 NR_040769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020577.mm.1 TC0900002458.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020577.mm.1 NR_040769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020577.mm.1 NR_040769.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020578.mm.1 TC0900002458.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020578.mm.1 NR_040769.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020578.mm.1 NR_040769.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0