Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936411_m1
          See other F730043M19RIK GE Assays ›
          Gene Symbol:
          F730043M19Rik
          Gene Name:
          RIKEN cDNA F730043M19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 33111711 - 33147586 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936411_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F730043M19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015602.2 5 - 6 1119 64
          GenBank mRNA AK043182.1 - 5 - 6 1120 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB255064.2 - 2 - 3 277 Chr. 12 - 33111716 - 33136032 [-] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cadherin-related family member 3 Cdhr3 1110049B09Rik,
          BB172694,
          Cdh28
          -18.7kb Sense Chr. 12 - 33033796 - 33093037 on NCBI Build GRCm38
          Downstream ataxin 7-like 1 Atxn7l1 2810423G08Rik,
          Atxn7l4
          28.3kb Anti-sense Chr. 12 - 33147639 - 33376408 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001616.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099910 AK043182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099910 NR_015602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P226348 AK162927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P226348 AK089514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P226348 AK162927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P226348 AK089514.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119797 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120643 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140514 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P226348 AK162927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P226348 AK089514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P226348 AK162927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P226348 AK089514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099910 AK043182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099910 NR_015602.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120643 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07581669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07581729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119797 - -
          Whole Mouse Genome G4122A A_52_P226348 AK162927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P226348 AK089514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P226348 AK162927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P226348 AK089514.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120017 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120643 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10084458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09856333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09809084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09860846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09769640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09898612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10384615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10123403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10115983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09976609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09976605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09996639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10186205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28339989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10336095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28340042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10399965 - -
          Affymetrix Mouse Clariom S Array PSR1200011264.mm.1 TC1200001616.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011264.mm.1 NR_015602.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 547051 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0