Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936437_s1
          See other 5730408K05RIK GE Assays ›
          Gene Symbol:
          5730408K05Rik
          Gene Name:
          RIKEN cDNA 5730408K05 gene
          Gene Aliases:
          2610018C13Rik
          Chromosome Location:
          Chr.19: 8888387 - 8888770 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936437_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5730408K05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027866.1 1 241 106
          GenBank mRNA AK017527.1 - 242 106
          AK019951.1 - 1 236 106
          AK043270.1 - 1 242 106
          BC027679.1 - 1 227 106
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV445307.1 - 1 207 Chr. 19 - 8888387 - 8888736 [-] on Build GRCm38 106
          GenBankmRNA BX637016.1 - 1 Chr. 19 - 8888387 - 8888755 [+] on Build GRCm38 106
          GenBankmRNA BY717316.1 - 1 240 Chr. 19 - 8887951 - 8888769 [-] on Build GRCm38 106
          GenBankmRNA DV041456.1 - 271 Chr. - - [-] on Build 106
          GenBankmRNA W41734.1 - 191 Chr. - - [-] on Build 106
          GenBankmRNA AA290313.1 - 1 214 Chr. 19 - 8888424 - 8888745 [-] on Build GRCm38 106
          GenBankmRNA BB257344.2 - 1 252 Chr. 19 - 8888390 - 8888770 [-] on Build GRCm38 106
          GenBankmRNA BB640694.1 - 1 240 Chr. 19 - 8888387 - 8888769 [-] on Build GRCm38 106
          GenBankmRNA AA939440.1 - 1 213 Chr. 19 - 8888387 - 8888745 [-] on Build GRCm38 106
          GenBankmRNA AA265907.1 - 1 155 Chr. 19 - 8888399 - 8888681 [-] on Build GRCm38 106
          GenBankmRNA BQ128601.1 - 1 Chr. 19 - 8888387 - 8888766 [+] on Build GRCm38 106
          GenBankmRNA BY717263.1 - 1 236 Chr. 19 - 8888387 - 8888764 [-] on Build GRCm38 106
          GenBankmRNA BQ128943.1 - 1 225 Chr. 19 - 8888389 - 8888754 [-] on Build GRCm38 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LBH domain containing 1 Lbhd1 Gm21743 Anti-sense Chr. 19 - 8883878 - 8890759 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1810009A15 gene 1810009A15Rik 113.0 Anti-sense Chr. 19 - 8888883 - 8890759 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001074.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2326582 AK017527.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465832 - -
          Affymetrix HT MG-430 PM Array Plate 1429208_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429207_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465831 - -
          Mouse Genome 430 2.0 Array 1429207_at - -
          Mouse Genome 430 2.0 Array 1429207_PM_at - -
          Mouse Genome 430 2.0 Array 1429208_PM_at - -
          Mouse Genome 430 2.0 Array 1429208_at - -
          Affymetrix Mouse Clariom S Array PSR1900010070.mm.1 TC1900001074.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010070.mm.1 NR_027866.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010070.mm.1 NR_027866.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010069.mm.1 TC1900001074.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010069.mm.1 NR_027866.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010069.mm.1 NR_027866.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010071.mm.1 TC1900001074.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010071.mm.1 NR_027866.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010071.mm.1 NR_027866.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010073.mm.1 TC1900001074.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900010073.mm.1 NR_027866.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010073.mm.1 NR_027866.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010073.mm.1 NR_039598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900010073.mm.1 NR_039598.1 Refseq
          Mouse Expression Set 430 1429207_PM_at - -
          Mouse Expression Set 430 1429208_at - -
          Mouse Expression Set 430 1429208_PM_at - -
          Mouse Expression Set 430 1429207_at - -
          Murine Genome U74v2 Set 104195_PM_at - -
          Murine Genome U74v2 Set 104195_at - -
          Mouse Genome 430A 2.0 Array 1429207_at - -
          Mouse Genome 430A 2.0 Array 1429208_at - -
          Mouse Genome 430A 2.0 Array 1429207_PM_at - -
          Mouse Genome 430A 2.0 Array 1429208_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429207_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429207_at - -
          Affymetrix HT Mouse Genome 430A Array 1429208_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429208_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102640450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0