Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03937065_s1
          See other E130317F20RIK GE Assays ›
          Gene Symbol:
          E130317F20Rik
          Gene Name:
          RIKEN cDNA E130317F20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 79851377 - 79854971 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03937065_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E130317F20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029447.1 1 1325 55
          GenBank mRNA AK045931.1 - 1 145 55
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB644479.1 - 1 145 Chr. 10 - 79853152 - 79853791 [-] on Build GRCm38 55

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitotic spindle positioning Misp 9130017N09Rik -21.9kb Anti-sense Chr. 10 - 79821019 - 79830452 on NCBI Build GRCm38
          Downstream polypyrimidine tract binding protein 1 Ptbp1 AA407203,
          AL033359,
          HNRPI,
          PTB-1,
          PTB2,
          PTB3,
          PTB4,
          Ptb,
          pPTB
          641.0 Anti-sense Chr. 10 - 79854432 - 79864435 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002481.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139504 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08662787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08375170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444729_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457912_PM_at - -
          Mouse Genome 430 2.0 Array 1444729_at - -
          Mouse Genome 430 2.0 Array 1457912_at - -
          Mouse Genome 430 2.0 Array 1444729_PM_at - -
          Mouse Genome 430 2.0 Array 1457912_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000018154.mm.1 TC1000002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018154.mm.1 NR_029447.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018154.mm.1 NR_029447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018152.mm.1 TC1000002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018152.mm.1 NR_029447.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018152.mm.1 NR_029447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018153.mm.1 TC1000002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018153.mm.1 NR_029447.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018153.mm.1 NR_029447.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018151.mm.1 TC1000002481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018151.mm.1 NR_029447.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018151.mm.1 NR_029447.1 Refseq
          Mouse Expression Set 430 1457912_at - -
          Mouse Expression Set 430 1457912_PM_at - -
          Mouse Expression Set 430 1444729_PM_at - -
          Mouse Expression Set 430 1444729_at - -
          Affymetrix HT Mouse Genome 430B Array 1457912_at - -
          Affymetrix HT Mouse Genome 430B Array 1444729_at - -
          Affymetrix HT Mouse Genome 430B Array 1457912_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444729_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0