Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03937079_s1
          See other B230327D02RIK GE Assays ›
          Gene Symbol:
          B230327D02Rik
          Gene Name:
          RIKEN cDNA B230327D02 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03937079_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230327D02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK045959.1 - 1 1754 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream numb gene homolog (Drosophila) Numb Nb Sense Chr. 12 - 83794034 - 83921934 on NCBI Build GRCm38
          Downstream RIKEN cDNA 2410016O06 gene 2410016O06Rik MAPJD,
          NO66
          35.3kb Anti-sense Chr. 12 - 83950608 - 83952953 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10322482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10084139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09970496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28595343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10425807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix