Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03937164_m1
          See other AW495222 GE Assays ›
          Gene Symbol:
          AW495222
          Gene Name:
          expressed sequence AW495222
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 93706100 - 93719175 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03937164_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AW495222
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045086.1 2 - 3 530 98
          GenBank mRNA AK046172.1 - 2 - 3 531 98
          BB314344.2 - 2 - 3 353 98
          BB644732.1 - 2 - 3 528 98
          BE656108.1 - 2 - 3 413 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY712700.1 - 2 - 3 502 Chr. 13 - 93706282 - 93719146 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream betaine-homocysteine methyltransferase 2 Bhmt2 C81077,
          D13Ucla2
          -31.9kb Sense Chr. 13 - 93656097 - 93674302 on NCBI Build GRCm38
          Downstream arylsulfatase B Arsb 1110007C02Rik,
          AI480648,
          As-1,
          As-1r,
          As-1s,
          As-1t,
          As1,
          As1-r,
          As1-s,
          As1-t,
          Asr-1,
          Ast-1
          55.4kb Anti-sense Chr. 13 - 93771023 - 93943016 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002438.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2273518 AK046172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2273518 AK046172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226499 AK046172.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126276 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444259_PM_at - -
          Mouse Genome 430 2.0 Array 1444259_PM_at - -
          Mouse Genome 430 2.0 Array 1444259_at - -
          Affymetrix Mouse Clariom S Array PSR1300016280.mm.1 TC1300002438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016280.mm.1 NR_045086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016280.mm.1 NR_045086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016285.mm.1 TC1300002438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016285.mm.1 NR_045086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016285.mm.1 NR_045086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016281.mm.1 TC1300002438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016281.mm.1 NR_045086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016281.mm.1 NR_045086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016283.mm.1 TC1300002438.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016283.mm.1 NR_045086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016283.mm.1 NR_045086.1 Refseq
          Mouse Expression Set 430 1444259_at - -
          Mouse Expression Set 430 1444259_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444259_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444259_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 541680 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline