Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03937623_g1
          See other B930082K07RIK GE Assays ›
          Gene Symbol:
          B930082K07Rik
          Gene Name:
          RIKEN cDNA B930082K07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 60687812 - 60690400 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03937623_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B930082K07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK047520.1 - 1 - 2 187 89
          AK082262.1 - 1 - 2 329 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB383801.2 - 1 - 2 351 Chr. 9 - 60687583 - 60688444 [+] on Build GRCm38 89
          GenBankmRNA BB647282.1 - 1 - 2 187 Chr. 9 - 60687719 - 60688546 [+] on Build GRCm38 89
          GenBankmRNA BB650785.1 - 1 - 2 324 Chr. 9 - 60687581 - 60688372 [+] on Build GRCm38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 9230112J17 gene 9230112J17Rik -141.8kb Anti-sense Chr. 9 - 60523967 - 60545882 on NCBI Build GRCm38
          Downstream La ribonucleoprotein domain family,
          member 6
          Larp6 5430431G03Rik,
          AI552438,
          Achn
          22.7kb Sense Chr. 9 - 60713121 - 60738801 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151398 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133200 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110875 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26560443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07662299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26560440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26560442 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 703797 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102470427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline