Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03938385_g1
          See other 2610037D02RIK GE Assays ›
          Gene Symbol:
          2610037D02Rik
          Gene Name:
          RIKEN cDNA 2610037D02 gene
          Gene Aliases:
          AU024679
          Chromosome Location:
          Chr.15: 96190474 - 96197974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03938385_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610037D02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040423.1 5 - 6 1211 79
          GenBank mRNA AK049239.1 - 5 - 6 1212 79
          AK163711.1 - 1425 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eukaryotic translation initiation factor 3,
          subunit I pseudogene
          Gm4371 EG667874,
          Gm8854
          Anti-sense Chr. 15 - 96182800 - 96203470 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4833422M21 gene 4833422M21Rik 4933406G12Rik 37.9kb Sense Chr. 15 - 96231870 - 96254684 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2256690 BB399856.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2256690 AK049239.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 650396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626489 NM_146266.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626489 NM_207008.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669474 AB079604.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669474 AF169152.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669474 AF169153.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669474 AK141231.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669474 NM_013879.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113732 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113732 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08470799 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08471039 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03107900 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03107971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08470865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08470829 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03107881 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03107916 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08470772 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113732 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08470799 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131814 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03107881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30461883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12421893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30461886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11963882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12080979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30461888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12080984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30461884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12080975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30462318 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453519_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459189_PM_at - -
          Mouse Genome 430 2.0 Array 1459189_at - -
          Mouse Genome 430 2.0 Array 1453519_PM_at - -
          Mouse Genome 430 2.0 Array 1453519_at - -
          Mouse Genome 430 2.0 Array 1459189_PM_at - -
          Mouse Expression Set 430 1453519_PM_at - -
          Mouse Expression Set 430 1453519_at - -
          Mouse Expression Set 430 1459189_PM_at - -
          Mouse Expression Set 430 1459189_at - -
          Murine Genome U74v2 Set 171556_PM_at - -
          Murine Genome U74v2 Set 171556_at - -
          Affymetrix HT Mouse Genome 430B Array 1459189_at - -
          Affymetrix HT Mouse Genome 430B Array 1453519_at - -
          Affymetrix HT Mouse Genome 430B Array 1453519_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459189_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 453598 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0