Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03938615_g1
          See other 4933409F18RIK GE Assays ›
          Gene Symbol:
          4933409F18Rik
          Gene Name:
          RIKEN cDNA 4933409F18 gene
          Gene Aliases:
          4930435E18Rik
          Chromosome Location:
          Chr.12: 26814375 - 26834888 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03938615_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933409F18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015322.1 - 5 - 6 1007 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700020D12 gene 1700020D12Rik -206.0kb Sense Chr. 12 - 26572113 - 26613757 on NCBI Build GRCm38
          Downstream predicted gene,
          20187
          Gm20187 269.6kb Sense Chr. 12 - 27090237 - 27114995 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2246269 AK015322.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467910 XM_001471910.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467910 XM_001471911.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11979 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02476635 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07567134 - -
          Whole Mouse Genome G4122A A_52_P236240 AK016753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236240 AK015322.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101056 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02476635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10399535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10018726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10319358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10059375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10018721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10319361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10187150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10041632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10015362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09904675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10129214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09919273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10015358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10399530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10196702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09847207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10225562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10361050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09919280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09919262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10369359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28304064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431555_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432171_PM_at - -
          Mouse Genome 430 2.0 Array 1431555_at - -
          Mouse Genome 430 2.0 Array 1432171_PM_at - -
          Mouse Genome 430 2.0 Array 1432171_at - -
          Mouse Genome 430 2.0 Array 1431555_PM_at - -
          Mouse Expression Set 430 1432171_PM_at - -
          Mouse Expression Set 430 1431555_PM_at - -
          Mouse Expression Set 430 1431555_at - -
          Mouse Expression Set 430 1432171_at - -
          Affymetrix HT Mouse Genome 430B Array 1432171_at - -
          Affymetrix HT Mouse Genome 430B Array 1431555_at - -
          Affymetrix HT Mouse Genome 430B Array 1431555_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432171_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102350369 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0