Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03938908_m1
          See other 4930480M12RIK GE Assays ›
          Gene Symbol:
          4930480M12Rik
          Gene Name:
          RIKEN cDNA 4930480M12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 26211267 - 26240707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03938908_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930480M12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046278.1 2 - 3 175 90
          GenBank mRNA AK015603.1 - 2 - 3 176 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715155.1 - 2 - 3 172 Chr. 12 - 26211270 - 26240707 [+] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          29968
          Gm29968 Anti-sense Chr. 12 - 26235530 - 26252548 on NCBI Build GRCm38
          Downstream ring finger protein 144A Rnf144a Rnf144,
          UIP4,
          Ubce7ip4
          66.1kb Anti-sense Chr. 12 - 26306794 - 26415321 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000223.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66199 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02475522 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123594 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10158831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09961701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10131745 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200001763.mm.1 TC1200000223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001763.mm.1 NR_046278.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001763.mm.1 NR_046278.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106020670 - -
          MouseWG-6 v2 Expression BeadChip 107050450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0