Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939159_s1
          See other 4930521G14RIK GE Assays ›
          Gene Symbol:
          4930521G14Rik
          Gene Name:
          RIKEN cDNA 4930521G14 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939159_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930521G14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015862.1 - 1 1246 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930544M13 gene 4930544M13Rik Sense Chr. 13 - 114607228 - 114697822 on NCBI Build GRCm38
          Downstream molybdenum cofactor synthesis 2 Mocs2 AI415403 181.4kb Sense Chr. 13 - 114818237 - 114829420 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2376188 AK015862.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29347460 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1460501_PM_at - -
          Mouse Genome 430 2.0 Array 1460501_at - -
          Mouse Genome 430 2.0 Array 1460501_PM_at - -
          Mouse Expression Set 430 1460501_at - -
          Mouse Expression Set 430 1460501_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460501_at - -
          Affymetrix HT Mouse Genome 430B Array 1460501_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104210020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0