Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939215_m1
          See other 4930532M18RIK GE Assays ›
          Gene Symbol:
          4930532M18Rik
          Gene Name:
          RIKEN cDNA 4930532M18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 154255689 - 154270416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939215_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930532M18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_108050.1 2 - 3 152 98
          GenBank mRNA AK015950.1 - 2 - 3 154 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CN841950.1 - 2 - 3 387 Chr. 1 - 154257003 - 154270322 [+] on Build GRCm38 98
          GenBankmRNA BY715514.1 - 2 - 3 153 Chr. 1 - 154255689 - 154270418 [+] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930452N14 gene 4930452N14Rik -6.1kb Anti-sense Chr. 1 - 154241521 - 154256906 on NCBI Build GRCm38
          Downstream calcium channel,
          voltage-dependent,
          R type,
          alpha 1E subunit
          Cacna1e A430040I15,
          BII,
          Cach6,
          Cacnl1a6,
          Cav2.3,
          Cchra1,
          alpha1E
          120.4kb Anti-sense Chr. 1 - 154390806 - 154884782 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001393.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129388 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04313572 - -
          Whole Mouse Genome G4122A A_52_P842768 AK015950.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00359739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00848899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00032392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00982268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00064199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00805486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20731023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00813395 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100011517.mm.1 TC0100001393.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011517.mm.1 NR_108050.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 680923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0