Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939370_m1
          See other 4930552N02RIK GE Assays ›
          Gene Symbol:
          4930552N02Rik
          Gene Name:
          RIKEN cDNA 4930552N02 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 52814604 - 52828269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939370_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930552N02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040661.1 3 - 4 291 112
          GenBank mRNA AK016098.1 - 3 - 4 292 112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715700.1 - 3 - 4 292 Chr. 4 - 52814604 - 52827972 [+] on Build GRCm38 112

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream olfactory receptor 275 Olfr275 MOR262-2 Sense Chr. 4 - 52825399 - 52826358 on NCBI Build GRCm38
          Downstream olfactory receptor 273 Olfr273 MOR262-8 27.3kb Anti-sense Chr. 4 - 52855558 - 52856511 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000570.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2357751 AK016098.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118575 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2357751 AK016098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05237882 - -
          Whole Mouse Genome G4122A A_52_P657537 AK016098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05237882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22919511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03687094 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432223_PM_at - -
          Mouse Genome 430 2.0 Array 1432223_PM_at - -
          Mouse Genome 430 2.0 Array 1432223_at - -
          Affymetrix Mouse Clariom S Array PSR0400004166.mm.1 TC0400000570.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004166.mm.1 NR_040661.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004166.mm.1 NR_040661.1 Refseq
          Mouse Expression Set 430 1432223_PM_at - -
          Mouse Expression Set 430 1432223_at - -
          Affymetrix HT Mouse Genome 430B Array 1432223_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432223_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 599890 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101340156 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0