Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939456_s1
          See other 4930557A04RIK GE Assays ›
          Gene Symbol:
          4930557A04Rik
          Gene Name:
          RIKEN cDNA 4930557A04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 9849703 - 9850252 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939456_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930557A04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029229.1 NP_083505.1 1 449 78
          GenBank mRNA AK016155.1 - 1 450 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715758.1 - 1 448 Chr. X - 9849704 - 9850254 [+] on Build GRCm38 78
          GenBankmRNA BU962030.1 - 451 Chr. - - [-] on Build 78
          GenBankmRNA BX638821.1 - 1 Chr. X - 9850099 - 9850256 [-] on Build GRCm38 78
          GenBankmRNA BU961195.1 - 449 Chr. - - [-] on Build 78
          GenBankmRNA CN834460.1 - 1 446 Chr. X - 9849709 - 9850234 [+] on Build GRCm38 78
          GenBankmRNA CN834758.1 - 446 Chr. - - [-] on Build 78
          GenBankmRNA CA464528.1 - 447 Chr. - - [-] on Build 78
          GenBankmRNA BU939461.1 - 1 446 Chr. X - 9849709 - 9850278 [+] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream synaptotagmin-like 5 Sytl5 ENSMUSG00000054453,
          slp5
          Sense Chr. X - 9747557 - 9998864 on NCBI Build GRCm38
          Downstream sushi-repeat-containing protein Srpx drs,
          drs-1,
          drs-2,
          mDRS-1,
          mDRS-2
          187.7kb Anti-sense Chr. X - 10037977 - 10117661 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000121.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284067 NM_029229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284067 AK016155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284067 NM_029229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P284067 AK016155.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121936 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284067 NM_029229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284067 AK016155.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284067 NM_029229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P284067 AK016155.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121936 - -
          Whole Mouse Genome G4122A A_51_P284067 NM_029229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284067 AK016155.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284067 NM_029229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284067 AK016155.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431673_PM_at - -
          Mouse Genome 430 2.0 Array 1431673_PM_at - -
          Mouse Genome 430 2.0 Array 1431673_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 385317_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 385317_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000823.mm.1 TC0X00000121.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000823.mm.1 NM_029229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000823.mm.1 NM_029229.1 Refseq
          Mouse Expression Set 430 1431673_PM_at - -
          Mouse Expression Set 430 1431673_at - -
          Affymetrix HT Mouse Genome 430B Array 1431673_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431673_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 776877 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2340129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2678394 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2678394 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0