Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939507_s1
          See other 4930563N14RIK GE Assays ›
          Gene Symbol:
          4930563N14Rik
          Gene Name:
          RIKEN cDNA 4930563N14 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939507_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930563N14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016211.1 - 1 751 114

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cell division cycle 20B Cdc20b EG238896,
          EG622422
          Anti-sense Chr. 13 - 113032954 - 113121019 on NCBI Build GRCm38
          Downstream granzyme A Gzma AW494114,
          Ctla-3,
          Ctla3,
          Hf,
          SE1,
          TSP-1,
          TSP1
          14.7kb Sense Chr. 13 - 113093827 - 113100981 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10590651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11026717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10868228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10723341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10915121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10637118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29338565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433365_PM_at - -
          Mouse Genome 430 2.0 Array 1433365_PM_at - -
          Mouse Genome 430 2.0 Array 1454577_at - -
          Mouse Genome 430 2.0 Array 1454577_PM_at - -
          Mouse Genome 430 2.0 Array 1433365_at - -
          Mouse Expression Set 430 1454577_PM_at - -
          Mouse Expression Set 430 1433365_at - -
          Mouse Expression Set 430 1454577_at - -
          Mouse Expression Set 430 1433365_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433365_at - -
          Affymetrix HT Mouse Genome 430B Array 1433365_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 334688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0