Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939650_s1
          See other 4930590H14RIK GE Assays ›
          Gene Symbol:
          4930590H14Rik
          Gene Name:
          RIKEN cDNA 4930590H14 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939650_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930590H14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016379.1 - 1 155 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CN834921.1 - 157 Chr. - - [-] on Build 99
          GenBankmRNA BY716027.1 - 1 152 Chr. 9 - 77357355 - 77357902 [+] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream muscular LMNA-interacting protein Mlip 2310046A06Rik,
          AI931782,
          CIP
          -9.5kb Anti-sense Chr. 9 - 77102084 - 77347870 on NCBI Build GRCm38
          Downstream RIKEN cDNA 5730403I07 gene 5730403I07Rik 8.5kb Anti-sense Chr. 9 - 77366412 - 77399416 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08010325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07780872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07575163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08107747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08163602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07837015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26653867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08107746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0